Key Documents
C9152
Sigma-Aldrich
Coumarin 35
Synonym(s):
7-(Diethylamino)-4-trifluoromethylcoumarin
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C14H14F3NO2
CAS Number:
Molecular Weight:
285.26
EC Number:
MDL number:
UNSPSC Code:
12171500
SMILES string
CCN(CC)c1ccc2c(OC(=O)C=C2C(F)(F)F)c1
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Kato, K.
IEEE J. Quant. Elect., 373-373 (1975)