Key Documents
C6779
Sigma-Aldrich
L-Orn(Z)-OH
Synonym(s):
Nδ-Cbz-L-ornithine
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C13H18N2O4
CAS Number:
Molecular Weight:
266.29
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
storage temp.
−20°C
SMILES string
N[C@@H](CCCNC(=O)OCc1ccccc1)C(O)=O
Looking for similar products? Visit Product Comparison Guide
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)