Key Documents
C5299
Sigma-Aldrich
cis-3,5-Cyclohexadiene-1,2-diol
20 % (w/v) in ethyl acetate
Synonym(s):
cis-1,2-Dihydrocatechol, cis-Benzene-1,2-glycol
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C6H8O2
CAS Number:
Molecular Weight:
112.13
MDL number:
UNSPSC Code:
12162002
concentration
20 % (w/v) in ethyl acetate
storage temp.
−20°C
SMILES string
O[C@@H]1C=CC=C[C@@H]1O
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)