Key Documents
C1680
Sigma-Aldrich
Chromostatin-20
≥97% (HPLC)
Synonym(s):
Ser-Asp-Glu-Asp-Ser-Asp-Gly-Asp-Arg-Pro-Gln-Ala-Ser-Pro-Gly-Leu-Gly-Pro-Gly-Pro
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C78H120N24O35
CAS Number:
Molecular Weight:
1953.93
MDL number:
UNSPSC Code:
12352200
assay
≥97% (HPLC)
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... KREMEN1(83999)
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
E Galindo et al.
Proceedings of the National Academy of Sciences of the United States of America, 88(4), 1426-1430 (1991-02-15)
Chromogranin A (CGA) is a ubiquitous 48-kDa secretory protein present in adrenal medulla, anterior pituitary, central and peripheral nervous system, endocrine gut, thyroid, parathyroid, and endocrine pancreas. Recently, we have demonstrated that the protein could be a precursor of bioactive