Key Documents
C1109
Sigma-Aldrich
CYFRA 21-1 human
Synonym(s):
Cytokeratin 19 Fragment human
Sign Into View Organizational & Contract Pricing
About This Item
MDL number:
UNSPSC Code:
12352200
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... KRT19(3880)
Biochem/physiol Actions
Tumor marker useful for detection of several cancer types.
Physical form
Liquid tissue culture supernatant in PBS buffer, 0.1% NaN3.
Choose from one of the most recent versions:
Certificates of Analysis (COA)