Key Documents
B8150
Sigma-Aldrich
5-Bromo-2′-deoxyuridine-2-14C
aqueous ethanol solution
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C9H11BrN2O5
CAS Number:
Molecular Weight:
307.10
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
form
aqueous ethanol solution
extent of labeling
40-60 mCi per mmol
shipped in
dry ice
storage temp.
−20°C
SMILES string
OCC1OC(CC1O)N2C=C(Br)C(=O)NC2=O
Packaging
serum bottle
Physical form
Aqueous solution containing 2% ethanol
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.