Key Documents
B3559
Sigma-Aldrich
Biotinyl-Melanin-Concentrating Hormone from salmon
Synonym(s):
MCH Biotinylated
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C99H153N29O26S5
Molecular Weight:
2325.78
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
form
solid
solubility
water: soluble
shipped in
ambient
storage temp.
−20°C
SMILES string
CSCC[C@@H]1NC(=O)[C@H](CSSC[C@H](NC(=O)[C@@H]2CCCN2C(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](Cc3ccc(O)cc3)NC(=O)[C@@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)CNC(=O)[C@@H](NC1=O)C(C)C)C(C)C)C(=O)N[C@@H](Cc4c[nH]c5ccccc45)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](C(C)C)C(O)=O)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCSC)NC(=O)[C@@H](NC(=O)[C@H](CC(O)=O)NC(=O)CCCC[C@@H]6SC[C@@H]7NC(=O)N[C@H]67)[C@@H](C)O
InChI
1S/C99H153N29O26S5/c1-48(2)75-91(148)110-44-72(132)111-57(20-14-34-106-96(100)101)82(139)124-76(49(3)4)92(149)119-63(40-52-26-28-54(130)29-27-52)85(142)117-62(22-16-36-108-98(104)105)94(151)128-37-17-23-69(128)90(147)121-68(89(146)118-64(41-53-43-109-56-19-11-10-18-55(53)56)86(143)114-59(30-31-73(133)134)83(140)125-77(50(5)6)95(152)153)47-159-158-46-67(88(145)115-61(33-39-156-9)84(141)123-75)120-80(137)58(21-15-35-107-97(102)103)113-81(138)60(32-38-155-8)116-93(150)78(51(7)129)126-87(144)65(42-74(135)136)112-71(131)25-13-12-24-70-79-66(45-157-70)122-99(154)127-79/h10-11,18-19,26-29,43,48-51,57-70,75-79,109,129-130H,12-17,20-25,30-42,44-47H2,1-9H3,(H,110,148)(H,111,132)(H,112,131)(H,113,138)(H,114,143)(H,115,145)(H,116,150)(H,117,142)(H,118,146)(H,119,149)(H,120,137)(H,121,147)(H,123,141)(H,124,139)(H,125,140)(H,126,144)(H,133,134)(H,135,136)(H,152,153)(H4,100,101,106)(H4,102,103,107)(H4,104,105,108)(H2,122,127,154)/t51-,57+,58+,59+,60+,61+,62+,63+,64+,65+,66+,67+,68+,69+,70+,75+,76+,77+,78+,79+/m1/s1
InChI key
FKLSNVGEODBRRJ-KJQIIMNASA-N
Amino Acid Sequence
Biotinyl-Asp-Thr-Met-Arg-Cys-Met-Val-Gly-Arg-Val-Tyr-Arg-Pro-Cys-Trp-Glu-Val-OH [Disulfide bridge: 5-14]
Biochem/physiol Actions
Originally isolated from salmon pituitary glands, also found in the mammalian CNS. In lower vertebrates, MCH alters pigmentation by inducing melanosome aggregation within melanocytes of teleost fish, whereas it causes melanosome dispersion in amphibians and reptiles.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.