Key Documents
B1754
Sigma-Aldrich
N-Benzoyl-DL-methionine
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C12H15NO3S
CAS Number:
Molecular Weight:
253.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
storage temp.
−20°C
SMILES string
CSCCC(NC(=O)c1ccccc1)C(O)=O
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)