Key Documents
B1162
Sigma-Aldrich
Brevetoxin 9
Synonym(s):
PbTx-9
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C50H74O14
CAS Number:
Molecular Weight:
899.11
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
storage temp.
−20°C
SMILES string
CC(CO)C1CC(O)C2(C)OC3CC4OC5CC6(C)OC7(C)CCC8OC9CC%10(C)OC%11C(CC%10OC9CC(C)C8OC7CC6OC5(C)CC=CC4OC3CC2O1)OC(=O)C=C%11C
General description
Polycyclic polyethers from the marine dinoflagellate Ptychodiscus brevis (Gymnodinium breve).
Biochem/physiol Actions
Potent toxins responsible for "red tide" fish kills, these compounds disrupt neurotransmission by opening sodium channels.
Other Notes
Contains a propanol side-chain.
Analysis Note
Semisynthetic, produced from Brevetoxin 3
pictograms
Skull and crossbonesEnvironment
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 1 Oral - Aquatic Acute 1 - Aquatic Chronic 1
Storage Class
6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Haiyan Wu et al.
Journal of chromatography. A, 1358, 172-180 (2014-08-05)
A liquid chromatography quadrupole linear ion trap mass spectrometry method with fast polarity switching and a scheduled multiple reaction monitoring algorithm mode was developed for multiclass screening and identification of lipophilic marine biotoxins in bivalve molluscs. A major advantage of