About This Item
Empirical Formula (Hill Notation):
C50H71N13O12
CAS Number:
Molecular Weight:
1046.18
MDL number:
UNSPSC Code:
12352200
assay
≥97% (HPLC)
storage temp.
−20°C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
F Rioux et al.
Canadian journal of physiology and pharmacology, 53(3), 383-391 (1975-06-01)
The C-terminal group of angiotensin II (ATii) , 1-Sar-ATii, and 1-beta-Asp-ATii was esterified to reduce degradation of the peptides and carboxypeptidases. Biological activity of esterified angiotensins was measured in vivo (rat blood pressure) and in vitro (rabbit aorta strip). Degradation