Key Documents
A6916
Sigma-Aldrich
Atrial Natriuretic Peptide fragment 4-28 human
≥97% (HPLC)
Synonym(s):
Auriculin® B, hANF 4-28
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C112H175N39O35S3
CAS Number:
Molecular Weight:
2724.02
MDL number:
UNSPSC Code:
12352200
assay
≥97% (HPLC)
UniProt accession no.
storage temp.
−20°C
SMILES string
CC[C@H](C)[C@@H]1NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCSC)NC(=O)[C@H](CCCNC(N)=N)NC(=O)CNC(=O)CNC(=O)[C@H](Cc2ccccc2)NC(=O)[C@H](CSSC[C@H](NC(=O)CNC(=O)[C@H](CC(C)C)NC(=O)CNC(=O)[C@H](CO)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](C)NC(=O)CNC1=O)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CO)C(=O)N[C@@H](Cc3ccccc3)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](Cc4ccc(O)cc4)C(O)=O)NC(=O)[C@H](CO)NC(=O)[C@H](CO)NC(=O)[C@@H](N)CCCNC(N)=N
Gene Information
human ... NPPA(4878)
application
Useful as an immunogen for producing antibodies to hANF.
Biochem/physiol Actions
Atrial natriuretic peptide (ANP) is synthesized in myoendocrine cells of the heart from which it is released into the circulation. It exerts natriuretic, diuretic, and vasodilatory effects through stimulation of guanylate cyclase-linked NPR-A receptors. It plays an important role in blood volume and blood pressure homeostasis.
Legal Information
Auriculin is a registered trademark of California Biotechnology, Inc.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
L S Weisberg et al.
Archives of internal medicine, 157(16), 1833-1838 (1997-09-18)
Acute tubular necrosis (ATN) is the most common type of acute renal failure in hospitalized patients and is associated with a high morbidity and mortality. The cause of ATN can be divided into nephrotoxic, ischemic, or mixed. To test the
K C Lee et al.
American journal of hypertension, 3(4), 262-267 (1990-04-01)
The renal and hemodynamic responses to intravenous anaritide, a human atrial natriuretic factor [102-126] at 0.3 to 20 micrograms/kg in conscious rhesus monkeys with and without acute extracellular hypervolemia were analyzed and compared. Acute isotonic saline loading (intravenous bolus at
Acute renal failure--the promise of new therapies.
H D Humes
The New England journal of medicine, 336(12), 870-871 (1997-03-20)
A B Weder et al.
Hypertension (Dallas, Tex. : 1979), 10(6), 582-589 (1987-12-01)
Synthetic atrial natriuretic factor (ANF) was administered in ascending doses (0.03, 0.20, 0.45 microgram/kg/min) to eight mildly essential hypertensive men on high (200 mEq/day) or low (10 mEq/day) sodium diets. Responses of blood pressure, heart rate, urinary volume and electrolyte
T A Fried et al.
The American journal of physiology, 255(2 Pt 2), F313-F316 (1988-08-01)
With micropuncture techniques, the present study examined the delivery of chloride to the superficial late distal tubule and the base and tip of the papillary collecting duct in rats treated with either Wy 47663, a synthetic analogue of atrial natriuretic