Key Documents
A6845
Sigma-Aldrich
N-Acetyl-Trp-Glu-His-Asp-p-nitroanilide
≥98% (HPLC), powder
Sign Into View Organizational & Contract Pricing
About This Item
MDL number:
UNSPSC Code:
12352200
assay
≥98% (HPLC)
form
powder
color
light yellow
solubility
DMSO: 20 mM
storage temp.
−20°C
Biochem/physiol Actions
Colorimetric substrate for caspase 1 and 5.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)