About This Item
Empirical Formula (Hill Notation):
C8H12N2O7
CAS Number:
Molecular Weight:
248.19
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
storage temp.
−20°C
SMILES string
N[C@@H](CC(O)=O)C(=O)N[C@@H](CC(O)=O)C(O)=O
Gene Information
human ... CA1(759) , CA2(760) , SLC15A1(6564)
rat ... Folh1(85309)
Choose from one of the most recent versions:
Already Own This Product?