Key Documents
A5286
Sigma-Aldrich
7-Amino-4-hydroxy-2-naphthalenesulfonic acid
powder
Synonym(s):
6-Amino-1-naphthol-3-sulfonic acid, I-Acid
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C10H9NO4S
CAS Number:
Molecular Weight:
239.25
Beilstein/REAXYS Number:
2217192
EC Number:
MDL number:
UNSPSC Code:
12352103
form
powder
color
yellow
SMILES string
Nc1ccc2c(O)cc(cc2c1)S(O)(=O)=O
InChI
1S/C10H9NO4S/c11-7-1-2-9-6(3-7)4-8(5-10(9)12)16(13,14)15/h1-5,12H,11H2,(H,13,14,15)
InChI key
KYARBIJYVGJZLB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Rino Ragno et al.
Journal of medicinal chemistry, 50(6), 1241-1253 (2007-02-28)
The screening of the inhibition capabilities of dye-like small molecules from a focused library against both human PRMT1 and Aspergillus nidulans RmtA is reported as well as molecular modeling studies (homology modeling, molecular docking, and 3-D QSAR) of the catalytic