Key Documents
A5183
Sigma-Aldrich
Ala-Pro-Arg-Leu-Arg-Phe-Tyr-Ser
≥97% (HPLC)
Synonym(s):
α-Bag cell peptide 1-8
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C47H72N14O11
CAS Number:
Molecular Weight:
1009.16
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
assay
≥97% (HPLC)
composition
Peptide content, ≥60%
storage temp.
−20°C
SMILES string
CC(C)CC(NC(=O)C(CCCNC(N)=N)NC(=O)C1CCCN1C(=O)C(C)N)C(=O)NC(CCCNC(N)=N)C(=O)NC(Cc2ccccc2)C(=O)NC(Cc3ccc(O)cc3)C(=O)NC(CO)C(O)=O
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
S M Pulst et al.
Neuroscience letters, 70(1), 40-45 (1986-09-25)
Double-label immunocytochemistry reveals that immunoreactivity for two putative peptide transmitters, egg-laying hormone and alpha-bag cell peptide, co-exist in most bag cell somata and processes in the abdominal ganglion of the marine mollusc Aplysia. Together with previous physiological and biochemical data
M K Rock et al.
Journal of neurobiology, 17(4), 273-290 (1986-07-01)
We have examined the effects of peptides on the neuroendocrine bag cells, the R2 neuron and the left upper quadrant (LUQ) neurons of the abdominal ganglion of Aplysia californica. Peptides include those extracted from the atrial gland, a reproductive organ;
S M Pulst et al.
Neuroscience, 27(1), 363-371 (1988-10-01)
Egg-laying hormone and alpha-bag cell peptide are two neuropeptides derived from a common precursor protein in the marine mollusk Aplysia. Previous studies indicate that they are neurotransmitters that co-exist in individual bag cell neurons and most bag cell processes in
S M Pulst et al.
Neuropeptides, 10(3), 249-259 (1987-10-01)
alpha-bag cell peptide (alpha BCP) is a putative neurotransmitter released from bag cell neurons of the marine mollusc Aplysia. alpha BCP is present in bag cell extracts and releasate from bag cells in two neuroactive forms: alpha BCP[1-9] and alpha
B S Rothman et al.
Proceedings of the National Academy of Sciences of the United States of America, 80(18), 5753-5757 (1983-09-01)
A discharge of impulse activity in a group of neuroendocrine cells, the bag cells, produces several types of prolonged responses in various identified neurons of the abdominal ganglion of Aplysia. Two excitatory responses are almost certainly mediated by egg-laying hormone