Key Documents
A2292
Sigma-Aldrich
MN Glycan
≥90%
Synonym(s):
β-D-Man-(1→4)-D-GlcNAc, 2-Acetamido-2-deoxy-4-0-(β-D-mannopyranosyl)-D-glucopyranose
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C14H25NO11
CAS Number:
Molecular Weight:
383.35
MDL number:
UNSPSC Code:
12352201
assay
≥90%
storage temp.
2-8°C
Choose from one of the most recent versions:
Certificates of Analysis (COA)