Key Documents
A1128
Sigma-Aldrich
D-Ala-Gly-Gly
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C7H13N3O4
CAS Number:
Molecular Weight:
203.20
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
storage temp.
−20°C
SMILES string
CC(N)C(=O)NCC(=O)NCC(O)=O
Choose from one of the most recent versions:
Certificates of Analysis (COA)