Key Documents
A0895
Sigma-Aldrich
Adenosine-5′-carboxylic acid
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C10H11N5O5
CAS Number:
Molecular Weight:
281.22
MDL number:
UNSPSC Code:
12352204
storage temp.
−20°C
SMILES string
Nc1ncnc2n(cnc12)C3OC(C(O)C3O)C(O)=O
Choose from one of the most recent versions:
Certificates of Analysis (COA)