Key Documents
A0216
Sigma-Aldrich
N-Acetyl-Trp-Glu-His-Asp-7-amido-4-methylcoumarin
≥97% (HPLC), powder
Synonym(s):
Ac-WEHD-AMC
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C38H40N8O11
Molecular Weight:
784.77
MDL number:
UNSPSC Code:
12352200
assay
≥97% (HPLC)
form
powder
color
white
storage temp.
−20°C
SMILES string
CC(=O)N[C@@H](Cc1c[nH]c2ccccc12)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](Cc3c[nH]cn3)C(=O)N[C@@H](CC(O)=O)C(=O)Nc4ccc5C(C)=CC(=O)Oc5c4
InChI
1S/C38H40N8O11/c1-19-11-34(52)57-31-14-22(7-8-24(19)31)43-36(54)30(15-33(50)51)46-38(56)29(13-23-17-39-18-41-23)45-35(53)27(9-10-32(48)49)44-37(55)28(42-20(2)47)12-21-16-40-26-6-4-3-5-25(21)26/h3-8,11,14,16-18,27-30,40H,9-10,12-13,15H2,1-2H3,(H,39,41)(H,42,47)(H,43,54)(H,44,55)(H,45,53)(H,46,56)(H,48,49)(H,50,51)/t27-,28-,29-,30-/m0/s1
InChI key
NONBZTYCHSGHGG-KRCBVYEFSA-N
Gene Information
human ... CASP1(834)
mouse ... CASP1(12362)
rat ... CASP1(25166)
Biochem/physiol Actions
Fluorogenic substrate for caspase 1.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Lakshmi Galam et al.
American journal of physiology. Lung cellular and molecular physiology, 310(6), L572-L581 (2016-01-10)
Increasing evidence shows that hyperoxia is a serious complication of oxygen therapy in acutely ill patients that causes excessive production of free radicals leading to hyperoxia-induced acute lung injury (HALI). Our previous studies have shown that P2X7 receptor activation is
T A Rano et al.
Chemistry & biology, 4(2), 149-155 (1997-02-01)
Interleukin-1beta converting enzyme (ICE/caspase-1) is the protease responsible for interleukin-1beta (IL-1beta) production in monocytes. It was the first member of a new cysteine protease family to be identified. Members of this family have functions in both inflammation and apoptosis. A