316229
3,3′,4,4′-Tetrachlorobiphenyl-UL-14C
~95% (Radiochemical Purity), toluene solution
About This Item
assay
~95% (Radiochemical Purity)
form
toluene solution
extent of labeling
1-25 mCi per mmol
storage temp.
2-8°C
Packaging
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.