Key Documents
314161
Sigma-Aldrich
2-Propanol-2-14C
Synonym(s):
Isopropanol
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C3H8O
CAS Number:
Molecular Weight:
60.10
MDL number:
UNSPSC Code:
12352200
extent of labeling
1-10 mCi per mmol
storage temp.
2-8°C
Packaging
Break-seal ampule
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.