About This Item
assay
≥98% (GC, HPLC)
extent of labeling
1-15 mCi per mmol
format
neat
storage temp.
2-8°C
SMILES string
O[14CH2]c1ccccc1
Looking for similar products? Visit Product Comparison Guide
Packaging
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.