Key Documents
403032
Sigma-Aldrich
Sodium arsenate dibasic heptahydrate
ACS reagent, ≥98%
Synonym(s):
Disodium hydrogen arsenate heptahydrate, Sodium hydrogenarsenate heptahydrate
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
Na2HAsO4 · 7H2O
CAS Number:
Molecular Weight:
312.01
MDL number:
UNSPSC Code:
12352300
grade
ACS reagent
assay
≥98%
98.0-102.0% (ACS specification)
impurities
≤0.005% insolubles
≤0.01% As2O3
mp
180 °C (dec.) (lit.)
anion traces
chloride (Cl-): ≤0.001%
nitrate (NO3-): ≤0.005%
sulfate (SO42-): ≤0.01%
cation traces
Fe: ≤0.001%
heavy metals (as Pb): ≤0.002%
SMILES string
[Na+].[Na+].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].O[As]([O-])([O-])=O
InChI
1S/AsH3O4.2Na.7H2O/c2-1(3,4)5;;;;;;;;;/h(H3,2,3,4,5);;;7*1H2/q;2*+1;;;;;;;/p-2
InChI key
KOLXPEJIBITWIQ-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A
Storage Class
6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Shugo Suzuki et al.
Toxicology, 299(2-3), 155-159 (2012-06-06)
Inorganic arsenic is a known human carcinogen, inducing tumors of the skin, urinary bladder and lung. It is metabolized to organic methylated arsenicals. 2,3-Dimercaptopropane-1-sulfonic acid (DMPS), a chelating agent, is capable of reducing pentavalent arsenicals to the trivalent state and
Tatyana S Pinyayev et al.
Chemical research in toxicology, 24(4), 475-477 (2011-03-11)
The conventional scheme for arsenic methylation accounts for methylated oxyarsenical production but not for thioarsenical formation. Here, we report that in vitro anaerobic microbiota of mouse cecum converts arsenate into oxy- and thio- arsenicals. Besides methylarsonic acid (MMA(V)), arsenate was
Dae Hyun Baek et al.
Journal of applied toxicology : JAT, 32(8), 617-626 (2011-12-02)
The embryonic stem cell test (EST) is a validated in vitro embryotoxicity test; however, as the inhibition of cardiac differentiation alone is used as a differentiation endpoint in the EST, it may not be a useful test to screen embryotoxic
Deniz Yildiz et al.
Journal of trace elements in medicine and biology : organ of the Society for Minerals and Trace Elements (GMS), 26(1), 53-58 (2011-12-20)
The objective of the present study was to investigate if arsenate V exposure results in glutathione efflux from human erythrocytes. The changes in intracellular and extracellular nonprotein sulfhydryl and glutathione levels were determined in arsenate (V) exposed erythrocytes. Presence of
William Brattin et al.
Journal of toxicology and environmental health. Part A, 76(7), 449-457 (2013-04-25)
This study describes a method for measuring the relative oral bioavailability (RBA) of arsenic (As) in soil and other soil-like media using young swine as the animal model. Groups of animals are exposed to site soil or sodium arsenate orally