Key Documents
402958
Sigma-Aldrich
Lithium tetraborate
ACS reagent, ≥98%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
Li2B4O7
CAS Number:
Molecular Weight:
169.12
EC Number:
MDL number:
UNSPSC Code:
12352300
grade
ACS reagent
assay
≥98%
98.0-102.0%
impurities
≤0.01% insolubles
loss
≤2.0% loss on fusion, 950°C
mp
760-880 °C (lit.)
density
≥0.25 g/mL at 25 °C (lit.)
cation traces
Al: ≤0.001%
Ca: ≤0.01%
Fe: ≤0.001%
K: ≤0.005%
Mg: ≤5 ppm
Na: ≤0.005%
Si: ≤0.01%
heavy metals: ≤0.001%
SMILES string
[Li+].[Li+].[O-]B(OB=O)OB([O-])OB=O
InChI
1S/B4O7.2Li/c5-1-9-3(7)11-4(8)10-2-6;;/q-2;2*+1
InChI key
PSHMSSXLYVAENJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)