Key Documents
W375306
Sigma-Aldrich
o-Tolyl isobutyrate
≥98%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
(CH3)2CHCO2C6H4CH3
CAS Number:
Molecular Weight:
178.23
FEMA Number:
3753
Council of Europe no.:
681
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.48
assay
≥98%
refractive index
n20/D 1.486 (lit.)
bp
107-108 °C/8 mmHg (lit.)
density
1.004 g/mL at 25 °C (lit.)
SMILES string
CC(C)C(=O)Oc1ccccc1C
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
203.0 °F - closed cup
flash_point_c
95.00 °C - closed cup
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)