Key Documents
W333018
Sigma-Aldrich
Benzyl trans-2-methyl-2-butenoate
≥90%
Synonym(s):
Benzyl tiglate
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
CH3CH=C(CH3)CO2CH2C6H5
CAS Number:
Molecular Weight:
190.24
FEMA Number:
3330
EC Number:
Council of Europe no.:
2184
MDL number:
UNSPSC Code:
12164502
Flavis number:
9.494
grade
Kosher
assay
≥90%
impurities
<10% benzyl ether
refractive index
n20/D 1.522 (lit.)
bp
139 °C/9 mmHg (lit.)
density
1.031 g/mL at 25 °C (lit.)
SMILES string
C\C=C(/C)C(=O)OCc1ccccc1
pictograms
Exclamation mark
signalword
Warning
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 2
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)