Key Documents
W286214
Sigma-Aldrich
Phenethyl isobutyrate
natural, ≥98%, FCC
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
(CH3)2CHCO2CH2CH2C6H5
CAS Number:
Molecular Weight:
192.25
FEMA Number:
2862
EC Number:
Council of Europe no.:
0302
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.427
grade
natural
reg. compliance
FCC
assay
≥98%
refractive index
n20/D 1.4873 (lit.)
bp
250 °C (lit.)
density
0.988 g/mL at 25 °C (lit.)
SMILES string
CC(C)C(=O)OCCc1ccccc1
InChI
1S/C12H16O2/c1-10(2)12(13)14-9-8-11-6-4-3-5-7-11/h3-7,10H,8-9H2,1-2H3
InChI key
JDQVBGQWADMTAM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Natural Occurrence: Peppermint oil, Scotch spearmint oil, blue and other cheeses, beer cognac, rum, cider, olive oil, and glanga.
Biochem/physiol Actions
Taste at 1-10 ppm
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 2
flash_point_f
226.4 °F - closed cup
flash_point_c
108.00 °C - closed cup
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)