Key Documents
W240702
Sigma-Aldrich
Elemi Resin
natural
Sign Into View Organizational & Contract Pricing
About This Item
CAS Number:
FEMA Number:
2407
EC Number:
MDL number:
UNSPSC Code:
12164502
biological source
Canarium luzonicum
grade
Kosher
natural
reg. compliance
FDA 21 CFR 172.51
origin
France origin
mp
28-30 °C (lit.)
application(s)
flavors and fragrances
documentation
see Safety & Documentation for available documents
food allergen
no known allergens
Preparation Note
Extraction method: solvent extraction
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)