Key Documents
W203602
Sigma-Aldrich
Allyl nonanoate
≥97%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
CH3(CH2)7CO2CH2CH=CH2
CAS Number:
Molecular Weight:
198.30
FEMA Number:
2036
EC Number:
Council of Europe no.:
390
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.109
biological source
synthetic
grade
Kosher
assay
≥97%
refractive index
n20/D 1.434 (lit.)
bp
151 °C/50 mmHg (lit.)
density
0.878 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
SMILES string
CCCCCCCCC(=O)OCC=C
InChI
1S/C12H22O2/c1-3-5-6-7-8-9-10-12(13)14-11-4-2/h4H,2-3,5-11H2,1H3
InChI key
MFLWLDDOGSNSKO-UHFFFAOYSA-N
Packaging
Packaged in glass bottles
Other Notes
Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.
pictograms
Exclamation mark
signalword
Warning
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)