Key Documents
T84700
Sigma-Aldrich
Triphenylphosphonium cyclopentadienide
Synonym(s):
(Cyclopentadienylidene)triphenylphosphorane
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C23H19P
CAS Number:
Molecular Weight:
326.37
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
mp
>225 °C (dec.) (lit.)
SMILES string
c1ccc(cc1)P(c2ccccc2)(c3ccccc3)=C4C=CC=C4
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)