Key Documents
M40405
Sigma-Aldrich
Methyl cyclopropanecarboxylate
98%
Synonym(s):
MCPC
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
C3H5CO2CH3
CAS Number:
Molecular Weight:
100.12
Beilstein/REAXYS Number:
2039388
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
98%
refractive index
n20/D 1.419 (lit.)
bp
119 °C/764 mmHg (lit.)
density
0.985 g/mL at 25 °C (lit.)
SMILES string
COC(=O)C1CC1
InChI
1S/C5H8O2/c1-7-5(6)4-2-3-4/h4H,2-3H2,1H3
InChI key
PKAHQJNJPDVTDP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
pictograms
FlameSkull and crossbones
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
64.4 °F - closed cup
flash_point_c
18 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)