Key Documents
D88488
Sigma-Aldrich
1-Diethylamino-2-propanol
97%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
(CH3CH2)2NCH2CH(OH)CH3
CAS Number:
Molecular Weight:
131.22
Beilstein/REAXYS Number:
969221
EC Number:
MDL number:
UNSPSC Code:
12352103
assay
97%
refractive index
n20/D 1.425 (lit.)
bp
55-59 °C/13 mmHg (lit.)
mp
13.5 °C (lit.)
density
0.889 g/mL at 25 °C (lit.)
SMILES string
CCN(CC)CC(C)O
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)