Key Documents
C31392
Sigma-Aldrich
2-Chlorocinnamic acid, predominantly trans
99%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
ClC6H4CH=CHCO2H
CAS Number:
Molecular Weight:
182.60
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
99%
mp
208-210 °C (lit.)
SMILES string
OC(=O)\C=C\c1ccccc1Cl
Choose from one of the most recent versions:
Certificates of Analysis (COA)