Key Documents
634980
Sigma-Aldrich
3-Amino-4(3H)-quinazolinone
97%
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C8H7N3O
CAS Number:
Molecular Weight:
161.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
assay
97%
form
solid
mp
209-213 °C (lit.)
SMILES string
NN1C=Nc2ccccc2C1=O
InChI
1S/C8H7N3O/c9-11-5-10-7-4-2-1-3-6(7)8(11)12/h1-5H,9H2
InChI key
XZRJWCFKYOZVIH-UHFFFAOYSA-N
pictograms
Exclamation mark
signalword
Warning
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Questions
Reviews
No rating value
Active Filters