Key Documents
630071
Sigma-Aldrich
Tributyltin hydride-functionalized silica gel
Sign Into View Organizational & Contract Pricing
About This Item
MDL number:
UNSPSC Code:
12352200
pictograms
Exclamation mark
signalword
Warning
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)