Key Documents
593540
Sigma-Aldrich
2-Methoxy-5-octyloxy-1,4-xylenebis(triphenylphosphoniumbromide) hydrate
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C53H56Br2O2P2 · xH2O
Molecular Weight:
946.77 (anhydrous basis)
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
mp
181-183 °C (lit.)
SMILES string
O.[Br-].[Br-].CCCCCCCCOc1cc(C[P+](c2ccccc2)(c3ccccc3)c4ccccc4)c(OC)cc1C[P+](c5ccccc5)(c6ccccc6)c7ccccc7
Choose from one of the most recent versions:
Certificates of Analysis (COA)