Key Documents
589330
Sigma-Aldrich
(3,5-Difluorophenylethynyl)trimethylsilane
98%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
(CH3)3SiC≡CC6H3F2
CAS Number:
Molecular Weight:
210.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
assay
98%
refractive index
n20/D 1.4920 (lit.)
bp
200-201 °C (lit.)
density
0.995 g/mL at 25 °C (lit.)
SMILES string
C[Si](C)(C)C#Cc1cc(F)cc(F)c1
InChI
1S/C11H12F2Si/c1-14(2,3)5-4-9-6-10(12)8-11(13)7-9/h6-8H,1-3H3
InChI key
SJTKBXIQLYSMRR-UHFFFAOYSA-N
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
165.7 °F - closed cup
flash_point_c
74.25 °C - closed cup
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Questions
Reviews
No rating value
Active Filters