Key Documents
579343
Sigma-Aldrich
(R)-(+)-Dimethyl-2,2′-dihydroxy-1,1′-binaphthalene-3,3′-dicarboxylate
98%
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C24H18O6
CAS Number:
Molecular Weight:
402.40
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
assay
98%
optical activity
[α]20/D +135°, c = 1% in chloroform
mp
245-248 °C (lit.)
InChI
1S/C24H18O6/c1-29-23(27)17-11-13-7-3-5-9-15(13)19(21(17)25)20-16-10-6-4-8-14(16)12-18(22(20)26)24(28)30-2/h3-12,25-26H,1-2H3
InChI key
SCOGIYZPQMECLW-UHFFFAOYSA-N
pictograms
Exclamation mark
signalword
Warning
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)