Key Documents
575003
Sigma-Aldrich
Potassium bis(oxalato)cuprate(II) hydrate
99.999% trace metals basis
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
K2[Cu(C2O4)2] · xH2O
Molecular Weight:
317.78 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay
99.999% trace metals basis
form
powder
mp
260 °C (dec.) (lit.)
SMILES string
[K+].[K+].[H]O[H].[O-]C(=O)C(=O)O[Cu]OC(=O)C([O-])=O
InChI
1S/2C2H2O4.Cu.2K.H2O/c2*3-1(4)2(5)6;;;;/h2*(H,3,4)(H,5,6);;;;1H2/q;;+2;2*+1;/p-4
InChI key
BTJGEIXAARTWRJ-UHFFFAOYSA-J
pictograms
Exclamation mark
signalword
Warning
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)