Key Documents
574813
Sigma-Aldrich
Bis(ethylenediamine)nickel(II) chloride hydrate
99.999% trace metals basis
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C4H16Cl2N4Ni · xH2O
Molecular Weight:
249.80 (anhydrous basis)
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay
99.999% trace metals basis
form
solid
mp
>300 °C (lit.)
SMILES string
[H]O[H].Cl[Ni]Cl.NCCN.NCCN
InChI
1S/2C2H8N2.2ClH.Ni.H2O/c2*3-1-2-4;;;;/h2*1-4H2;2*1H;;1H2/q;;;;+2;/p-2
InChI key
JCYYALJEEILBFR-UHFFFAOYSA-L
pictograms
Health hazard
signalword
Danger
Hazard Classifications
Carc. 1B - Resp. Sens. 1 - Skin Sens. 1
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)