Key Documents
574465
Sigma-Aldrich
2′,4′-Diethoxyacetophenone
98%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
CH3COC6H5(OC2H5)2
CAS Number:
Molecular Weight:
208.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
98%
form
solid
mp
68-70 °C (lit.)
SMILES string
CCOc1ccc(C(C)=O)c(OCC)c1
InChI
1S/C12H16O3/c1-4-14-10-6-7-11(9(3)13)12(8-10)15-5-2/h6-8H,4-5H2,1-3H3
InChI key
HVMNBJVJMFQOOZ-UHFFFAOYSA-N
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Questions
Reviews
No rating value
Active Filters