Key Documents
572624
Sigma-Aldrich
4-Methylbenzhydryl alcohol, polymer-bound
100-200 mesh, extent of labeling: 1.6-2.0 mmol/g loading, 1 % cross-linked with divinylbenzene
Sign Into View Organizational & Contract Pricing
About This Item
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
crosslinking
1 % cross-linked with divinylbenzene
extent of labeling
1.6-2.0 mmol/g loading
particle size
100-200 mesh
Application
Benzhydryl alcohol resins are a less acid labile alternative to Rink acid resin. They are used to anchor mostly carboxylic acids and alcohols using the Fmoc protecting group strategy. The 4-methylbenzhydryl alcohol resin is a more acid labile resin than the benzhydryl alcohol resin.
Choose from one of the most recent versions:
Certificates of Analysis (COA)