Key Documents
572004
Sigma-Aldrich
Pentadecyl isocyanate
97%
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C16H31NO
CAS Number:
Molecular Weight:
253.42
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
97%
refractive index
n20/D 1.4470 (lit.)
bp
320-324 °C (lit.)
density
0.852 g/mL at 25 °C (lit.)
SMILES string
CCCCCCCCCCCCCCCN=C=O
InChI
1S/C16H31NO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-17-16-18/h2-15H2,1H3
InChI key
HULTVDSPXGVYBQ-UHFFFAOYSA-N
pictograms
Health hazardExclamation mark
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
target_organs
Respiratory system
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
>230.0 °F
flash_point_c
> 110 °C
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)