Key Documents
569119
Sigma-Aldrich
4-(Dichloromethyl)benzoyl chloride
95%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
Cl2CHC6H4C(O)Cl
CAS Number:
Molecular Weight:
223.48
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
95%
mp
50-55 °C (lit.)
SMILES string
ClC(Cl)c1ccc(cc1)C(Cl)=O
pictograms
Corrosion
signalword
Danger
hcodes
Hazard Classifications
Skin Corr. 1B
Storage Class
8B - Non-combustible corrosive hazardous materials
wgk_germany
WGK 3
flash_point_f
>230.0 °F - closed cup
flash_point_c
> 110 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)