Key Documents
566616
Sigma-Aldrich
Poly((4,4′-hexafluoroisopropylidene)diphthalic anhydride-alt-thionin)
Sign Into View Organizational & Contract Pricing
About This Item
MDL number:
UNSPSC Code:
12352103
mp
>400 °C (dec.) (lit.)
solubility
DMF: soluble
DMSO: soluble
NMP: soluble
concentrated sulfuric acid: soluble
methanol: insoluble
λmax
265 nm
Features and Benefits
Fluorescent dye-containing polyimide with good solubility characteristics.
Choose from one of the most recent versions:
Certificates of Analysis (COA)