Key Documents
558079
Sigma-Aldrich
(S)-(+)-4-Methyl-2-pentanol
98%, optical purity99%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
(CH3)2CHCH2CH(OH)CH3
CAS Number:
Molecular Weight:
102.17
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
assay
98%
optical activity
[α]20/D +18.0°, c = 1% in chloroform
optical purity
99%
refractive index
n20/D 1.4790 (lit.)
bp
138 °C (lit.)
density
0.802 g/mL at 25 °C (lit.)
SMILES string
CC(C)C[C@H](C)O
Choose from one of the most recent versions:
Certificates of Analysis (COA)