Key Documents
557145
Sigma-Aldrich
1,1,1-Trichloro-2,2-bis(4-hydroxyphenyl)ethane
97%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
Cl3CCH(C6H4OH)2
CAS Number:
Molecular Weight:
317.59
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
97%
mp
204-208 °C (lit.)
SMILES string
Oc1ccc(cc1)C(c2ccc(O)cc2)C(Cl)(Cl)Cl
Gene Information
mouse ... Esr1(13982)
rat ... Ar(24208)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Xiaohui Liu et al.
PloS one, 16(2), e0246583-e0246583 (2021-02-10)
We reported that bisphenol AF (BPAF) works as an agonist for estrogen receptor (ER) ERα but as an antagonist for ERβ. Similar results were observed for bisphenol E analogs (BPE-X) such as BPE-F, BPE-Cl, and BPE-Br, each consisting of a
Takahiro Masuya et al.
Scientific reports, 9(1), 9954-9954 (2019-07-11)
Bisphenol A (BPA) is used as an industrial raw material for polycarbonate plastics and epoxy resins; however, various concerns have been reported regarding its status as an endocrine-disrupting chemical. BPA interacts not only with oestrogen receptors (ERs) but constitutive androstane