Key Documents
556769
Sigma-Aldrich
4-Acetylamino-2-(diethylamino)anisole
97%
Synonym(s):
3′-(Diethylamino)-4′-methoxyacetanilide
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
CH3C(O)NHC6H3(OCH3)N(CH2CH3)2
CAS Number:
Molecular Weight:
236.31
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
97%
mp
93-97 °C (lit.)
SMILES string
CCN(CC)c1cc(NC(C)=O)ccc1OC
InChI
1S/C13H20N2O2/c1-5-15(6-2)12-9-11(14-10(3)16)7-8-13(12)17-4/h7-9H,5-6H2,1-4H3,(H,14,16)
InChI key
BTJCIVXKBILNPY-UHFFFAOYSA-N
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)