Key Documents
553948
Sigma-Aldrich
Bis(methylcyclopentadienyl)niobium(IV) dichloride
97%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
(C5H4CH3)2NbCl2
CAS Number:
Molecular Weight:
322.05
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay
97%
mp
266-270 °C (lit.)
SMILES string
Cl[Nb]Cl.C[C]1[CH][CH][CH][CH]1.C[C]2[CH][CH][CH][CH]2
Choose from one of the most recent versions:
Certificates of Analysis (COA)