MilliporeSigma
All Photos(2)
Key Documents
546445
Sigma-Aldrich
2,5-Bis(chloromethyl)-1-methoxy-4-(3′,7′-dimethyloctyloxy)benzene
98%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
(ClCH2)2C6H2(OCH3)O(CH2)2CH(CH3)(CH2)3CH(CH3)2
CAS Number:
Molecular Weight:
361.35
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
assay
98%
mp
66-69 °C (lit.)
SMILES string
COc1cc(CCl)c(OCCC(C)CCCC(C)C)cc1CCl
InChI
1S/C19H30Cl2O2/c1-14(2)6-5-7-15(3)8-9-23-19-11-16(12-20)18(22-4)10-17(19)13-21/h10-11,14-15H,5-9,12-13H2,1-4H3
InChI key
NLOCNKYNECKGNC-UHFFFAOYSA-N
Application
Conducting polymer precursor for synthesis of substituted PPV.
pictograms
Corrosion
signalword
Danger
hcodes
Hazard Classifications
Skin Corr. 1B
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Questions
Reviews
No rating value
Active Filters